3,3'-Diaminobenzidine (DAB) 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:91-95-2
:
Formula:C₁₂H₁₄N₄
MDL Number:MFCD00007725
:214.27 g/mol
Melting Pt:176…180 °C
Flash Pt:>200 °C
EINECS:202-110-6
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Off-white to brown Powder or crystals
- Infrared spectrum:Conforms
- Melting point:175 to 179 °C
- HPLC:≥98.0 %