Order Entry
Germany
ContactUsLinkComponent
3,3'-Diaminobenzidine (DAB)
3,3'-Diaminobenzidine (DAB)
Catalog # MOLEM58703130
Supplier:  Molekula
CAS Number:  
3,3'-Diaminobenzidine (DAB)
Catalog # MOLEM58703130
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :214.27 g/mol
EINECS:202-110-6
CAS Number:91-95-2
Formula:C₁₂H₁₄N₄
MDL Number:MFCD00007725