(4-Methoxyphenyl)acetic acid ethyl ester
Catalog # ABCRAB138338.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
Specifications
About this item
:
:194.23 g/mol
Boiling Pt:138 °C (1013 hPa)
Density:1.097 g/cm³ (24 °C)
EINECS:237-903-6
CAS Number:14062-18-1
Formula:C₁₁H₁₄O₃
MDL Number:MFCD00040760
Boiling Pt:138 °C (1013 hPa)