(4-Methoxyphenyl)acetic acid ethyl ester 97%
:14062-18-1
:
:C₁₁H₁₄O₃
:MFCD00040760
:194.23 g/mol
:108…111 °C (3 mmHg)
:1.097 g/cm³
:46 °C
:237-903-6
:3272
:GHS02
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5065 to 1.5085 (20°C, 589 nm)
- GC:≥96.0 %