Acetonitrile ≥99.5% (by GC)
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:82 °C (1.013 hPa)
Melting Pt:–46 °C
Density:0.7857 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS08
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Refractive Index:n20/D 1.344 (lit.)
- Assay (GC):Min. 99.5 %
- Water (Karl Fischer):Max. 0.1 %
- Vapor Density:1.41 (vs air)
- Vapor Pressure:72.8 mmHg ( 20 °C)
- Explosion Limit:16 %