Order Entry
Germany
ContactUsLinkComponent
 
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
    
 
undefined
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Explosion Limit:
    16 %
  • Appearance:
    complying
  • Assay (GC):
    Min. 99.5 %
  • Boiling range:
    81 - 82 °C
  • Boiling range 80-82 °C:
    Min. 95 %
  • Density (D 20/4):
    0.781 - 0.783
  • Refractive index (n 20/D):
    1.3435 - 1.3445
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free acid (as CH₃COOH):
    Max. 0.001 %
  • Titrable Acid:
    Max. 8 µeq/g
  • Free alkali (as NH₃):
    Max. 0.001 %
  • Titrable Base:
    Max. 0.6 µeq/g
  • Aluminium (Al):
    Max. 0.00005 %
  • Boron (B):
    Max. 0.000002 %
  • Barium (Ba):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000002 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Copper (Cu):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.00001 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.00001 %
  • Tin (Sn):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.00001 %
  • APHA:
    Max. 10