Acetonitrile
Supplier: BIOLAB
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion