Acetonitrile 99.6% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:81…82 °C (760 mmHg)
Melting Pt:–46 °C
Density:0.781 g/cm³
Flash Pt:2 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.3 % (K.F.)
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
- Titrable acid:≤0.008 meq/g
- Titrable base:≤0.0006 meq/g