Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Catalog # 364310025.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.001 %
- GC:≥99.85 %
Specification Test Results
About this item
:
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:81…82 °C (760 mmHg)
Melting Pt:–46 °C
Density:0.781 g/cm³
Flash Pt:2 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS07
:GHS02
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878