ContactUsLinkComponent
Acetonitrile 99+%, extra pure
Acetonitrile 99+%, extra pure
   Sustainable Options Available
Certificates
undefined
Acetonitrile 99+%, extra pure
   Sustainable Options Available
Specifications
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
  • Appearance:
    Clear liquid
  • Water:
    ≤0.3 % (K.F.)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤20 APHA
  • Residue after evaporation:
    ≤0.005 %