Dispensers for KIMTECH* WETTASK* wipes
Catalog # 115-2076
Supplier: KIMBERLY CLARK
Specifications
- Description:Dispenser bucket, 4,5 l, blue
- Pk:4
- For:SXX/DSX wipes
Specifications
About this item
Dispenser buckets for KIMTECH* WETTASK* roll wipers. Bucket can easily be opened and refilled with wipes and sanitizers/solutions. This enclosed system reduces cross contamination, Volatile Organic Compounds and splashes.
- Easy refilling with the appropriate solution
- Pre-moistened cloths can easily be pulled through the aperture in the lid
- Mobile usage: Portable