Bromoacetaldehyde dimethyl acetal
Catalog # MOLE52286577-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:169.02 g/mol
Boiling Pt:148 °C (1013 hPa)
Density:1.43 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:230-669-6
UN:2929
ADR:6.1,II
:GHS02
:GHS07
CAS Number:7252-83-7
Formula:BrH₂CCH(OCH₃)₂
MDL Number:MFCD00000213