Bromoacetaldehyde dimethyl acetal
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:7252-83-7
:
Formula:BrH₂CCH(OCH₃)₂
MDL Number:MFCD00000213
:169.02 g/mol
Boiling Pt:148 °C (1013 hPa)
Density:1.43 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:230-669-6
UN:2929
ADR:6.1,II
:GHS02
:GHS07