
Bromotrichloromethane 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-62-7
:
Formula:BrCl₃C
MDL Number:MFCD00000783
:198.27 g/mol
Boiling Pt:105 °C (1013 hPa)
Melting Pt:–6 °C
Density:1.997 g/cm³
Storage Temperature:Ambient
EINECS:200-886-0
UN:2810
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5055 to 1.5075 (20°C, 589 nm)
- GC:≥98.5 %