
1,9-Dibromononane 97%
:4549-33-1
:
:BrH₂C(CH₂)₇CH₂Br
:MFCD00000279
:286.05 g/mol
:285 °C (1013 hPa)
:–10 °C
:1.40 g/cm³
:>112 °C
:224-913-0
:GHS07
:Ambient
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4950 to 1.4980 (20°C, 589 nm)
- GC:≥96.0 %
- Specific optical rotation:+39° to +69° (20°C, 589 nm) (C=2, Chloroform)