Order Entry
Germany
ContactUsLinkComponent
1,6-Dibromohexane for synthesis, Sigma-Aldrich®
Irritant
Environment
1,6-Dibromohexane for synthesis, Sigma-Aldrich®
Catalog # 8.02921.0250
Supplier:  Merck
CAS Number:  
Irritant
Environment
1,6-Dibromohexane for synthesis, Sigma-Aldrich®
Catalog # 8.02921.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    -85 °C
  • Molar Mass:
    243.96 g/mol
  • Boiling Point:
    243 °C
  • Vapor Pressure:
     - 1 hPa (20 °C)
  • Flash Point:
    110 °C
  • Refractive Index:
    1.5066 (20 °C, 589 nm)
  • Density:
    1.61 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :243.97 g/mol
Boiling Pt:243 °C (1013 hPa)
Melting Pt:–2 °C
Density:1.61 g/cm³ (20 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:211-067-2
UN:2810
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
 :GHS07
 :GHS09
CAS Number:629-03-8
Formula:BrH₂C(CH₂)₄CH₂Br
MDL Number:MFCD00000272