Order Entry
Germany
ContactUsLinkComponent
1,2-Dibromoethane for synthesis, Sigma-Aldrich®
Toxic
Health
Environment
1,2-Dibromoethane for synthesis, Sigma-Aldrich®
Catalog # 8.00952.1000
Supplier:  Merck
CAS Number:  
Toxic
Health
Environment
1,2-Dibromoethane for synthesis, Sigma-Aldrich®
Catalog # 8.00952.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, plastic-coated, returnable
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :187.86 g/mol
Merck Index:13,03830
Boiling Pt:131.7 °C (1013 hPa)
Melting Pt:10 °C
Density:2.18 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-444-5
UN:1605
ADR:6.1,I
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS08
 :GHS09
CAS Number:106-93-4
Formula:BrH₂CCH₂Br
MDL Number:MFCD00000233