ContactUsLinkComponent
Zinc(II) nitrate hexahydrate
Zinc(II) nitrate hexahydrate
  HONE14436.9025
 :  
Zinc(II) nitrate hexahydrate
  HONE14436.9025
 :  
  • Pk:
    25 kg
  • Seq:
    4283
 :
 :297.49 g/mol
 :12,10273
 :∼105 °C (1013 hPa)
 :36 °C
 :2.06 g/cm³ (20 °C)
 :Ambient
 :231-943-8
 :1514
 :5.1,II
 :GHS03
 :GHS07
 :10196-18-6
 :ZnN₂O₆·6H₂O
 :MFCD00149889