Lanthanum(III) nitrate hexahydrate 98+%
:10277-43-7
:10227-43-7
:La(NO₃)₃·6H₂O
:MFCD00149751
:433.01 g/mol
:14,05363
:126 °C (1013 hPa)
:40 °C
:2,347 g/cm³ (20 °C)
:Ambient
:233-238-0
:1477
:5.1,II
:GHS07
:GHS03
- Appearance:White Crystalline powder or crystals and/or chunks
- Titration Complexometric:≥98.0 %