Order Entry
Germany
ContactUsLinkComponent
Europium trinitrate hexahydrate 99.9% (trace metals basis)
Europium trinitrate hexahydrate 99.9% (trace metals basis)
Catalog # ACRO196680050
CAS Number:  
undefined
Europium trinitrate hexahydrate 99.9% (trace metals basis)
Catalog # ACRO196680050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Semitransparent Crystalline solid
  • Total conc. of imp.as REO:
    ≤0.1 %

Specification Test Results

About this item

 :
 :446.07 g/mol
Melting Pt:85 °C
Density:2,258 g/cm³ (20 °C)
EINECS:233-380-3
UN:1477
ADR:5.1,II
 :GHS03
 :GHS07
CAS Number:10031-53-5
Formula:EuN₃O₉·6H₂O
MDL Number:MFCD00149700