Order Entry
Germany
ContactUsLinkComponent
 
Tetrahydrofuran, anhydrous ≥99.8% stabilised
  44608.M1
 :  
undefined
Tetrahydrofuran, anhydrous ≥99.8% stabilised
  44608.M1
 :  

 

  • Pk:
    4
  • Seq:
    5049
  • Pk Info:
    4x1 L

 

 

 :
 :72.11 g/mol
 :14,9211
 :66 °C
 :–108 °C
 :0.889
 :–17 °C (1 °F)
 :3,II
 :Packaged under argon in resealable ChemSeal™ bottles.
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 0.025%
 :Ambient
 :203-726-8
 :2056
 :05-2114615188-48
 :GHS02
 :GHS08
 :GHS07
 :109-99-9
 :C₄H₈O
 :MFCD00005356