Order Entry
Germany
ContactUsLinkComponent
Aluminium chloride hexahydrate
Irritant
Aluminium chloride hexahydrate
Catalog # MOLE26726139-100G
Supplier:  Molekula
CAS Number:  
Irritant
Aluminium chloride hexahydrate
Catalog # MOLE26726139-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :241.43 g/mol
Melting Pt:∼100 °C
Density:2.39 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-208-1
UN:3260
ADR:8,III
 :GHS07
CAS Number:7784-13-6
Formula:AlCl₃·6H₂O
MDL Number:MFCD00149134