Diiodomethane ≥99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-11-6
:
Formula:CH₂I₂
MDL Number:MFCD00001079
:267.84 g/mol
Merck Index:14,6066
Boiling Pt:180…181 °C (dec.)
Melting Pt:5…6 °C
Density:3.322
Flash Pt:110 °C (closed cup)
UN:2810
ADR:6.1,III
:Stabilised with copper
EINECS:200-841-5
:GHS05
:GHS07
Storage Temperature:Ambient