Order Entry
Germany
ContactUsLinkComponent
Magnesium chloride hexahydrate 99.0-101.0% (by complexometric analysis), crystals, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JPC, FCC, E511, SAFC®
Magnesium chloride hexahydrate 99.0-101.0% (by complexometric analysis), crystals, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JPC, FCC, E511, SAFC®
Supplier:  Merck
Certificates
Magnesium chloride hexahydrate 99.0-101.0% (by complexometric analysis), crystals, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JPC, FCC, E511, SAFC®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (Complexometric):
    99.0 - 101.0%
  • Identity:
    Passes Test
  • Appearance of Solution:
    Passes Test
  • Insoluble Matter:
    ≤0.005%
  • Acidity or Alkalinity:
    Passes Test
  • pH-value (5%; Water):
    5.0 - 7.0
  • Bromide (Br):
    ≤0.05%
  • Sulfate (SO₄):
    ≤0.005%
  • Heavy Metals (as Pb):
    ≤0.001%
  • Al (Aluminium):
    ≤0.0001%
  • As (Arsenic):
    ≤0.0002%
  • Ba (Barium):
    Passes Test
  • Ca (Calcium):
    ≤0.01%
  • Fe (Iron):
    ≤0.0005%
  • Hg (Mercury):
    ≤0.0001%
  • K (Potassium):
    ≤0.0500%
  • NH₄ (Ammonium):
    ≤0.005%
  • Pb (Lead):
    ≤0.0002%
  • Residual Solvents (Ph. Eur./USP/ICH):
    Excluded by Manufacturing Process
  • Water:
    51.0 - 55.0%
  • Endotoxins:
    ≤3.0 I.U./g
  • Total Aerobic Microbial Count:
    ≤100 CFU/g
  • Total Yeast and Mould Count:
    ≤100 CFU/g

Specification Test Results

Related Information