Barium chloride dihydrate, Supelco®
: Merck
:10326-27-9
:
:BaCl₂·2H₂O
:MFCD00149154
:244.27 g/mol
:14,00971
:1560 °C (1013 hPa)
:960 °C
:3.86 g/cm³ (25 °C)
:1564
:6.1,III
:M-Clarity™ quality level = MQ200
:233-788-1
:GHS06
:Ambient