Order Entry
Germany
ContactUsLinkComponent
Disodium hydrogen phosphate heptahydrate
Disodium hydrogen phosphate heptahydrate
Catalog # HONE30413.9050
CAS Number:  
Disodium hydrogen phosphate heptahydrate
Catalog # HONE30413.9050
CAS Number:  

Specifications

  • Pk:
    50 kg
  • Seq:
    4418

Specifications

About this item

 :
 :268.07 g/mol
Melting Pt:48 °C
Density:1.7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-448-7
CAS Number:7782-85-6
Formula:HNa₂O₄P·7H₂O
MDL Number:MFCD00149180