
Sodium carbonate decahydrate Ph. Eur.
:6132-02-1
:
:Na₂CO₃·10H₂O
:MFCD00149178
:286.14 g/mol
:33 °C
:1.46 g/cm³ (20 °C)
:207-838-8
:GHS07
:Ambient
- Assay (Na2CO3):36.7 - 40.0 %
- Appearance Conforms:(see CoA/CoS)
- Identification A:Passes test
- Identification (CO3):Passes test
- Identification (Na):Passes test
- Appearance of solution:Passes test
- Alkali hydroxides and bicarbonates:Passes test
- Cl (Chloride):≤ 50 ppm
- SO4 (Sulphate):≤ 100 ppm
- Fe (Iron):≤ 20 ppm
- Heavy metals (as Pb):≤ 20 ppm
- Residual solvents:Passes test
- As (Arsenic):≤ 2 ppm
- Cd (Cadmium):≤ 1 ppm
- Co (Cobalt):≤ 1 ppm
- Hg (Mercury):≤ 3 ppm
- Ni (Nickel):≤ 1 ppm
- Pb (Lead):≤ 3 ppm
- V (Vanadium):≤ 1 ppm
- Conforms to Ph.Eur.:Passes test