
Sodium carbonate decahydrate analytical reagent
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:6132-02-1
:
Formula:Na₂CO₃·10H₂O
MDL Number:MFCD00149178
:286.14 g/mol
Melting Pt:33 °C
Density:1.46 g/cm³ (20 °C)
EINECS:207-838-8
:GHS07
Storage Temperature:Ambient