
Sodium carbonate decahydrate
Catalog # HONE31431.9025
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:286.14 g/mol
Melting Pt:33 °C
Density:1.46 g/cm³ (20 °C)
EINECS:207-838-8
:GHS07
Storage Temperature:Ambient
CAS Number:6132-02-1
Formula:Na₂CO₃·10H₂O
MDL Number:MFCD00149178