Order Entry
Germany
ContactUsLinkComponent
Magnesium chloride 0.1 mol/l (0.2 N) in aqueous solution, Sigma-Aldrich®
Magnesium chloride 0.1 mol/l (0.2 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIAL00457-1ML-F
Supplier:  Merck
CAS Number:  
Magnesium chloride 0.1 mol/l (0.2 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIAL00457-1ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :95.21 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106