
Sulphuric acid 0.25 mol/l (0.5 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Density:1.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:3264
ADR:8,III
:GHS05
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.2495 - 0.2505 mol/l
- NIST traceable:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Cubitainer UN (ZPACK13260004) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000)
Verpackungsinformationen
27417836.pdf spec_VWRC30143.291 spec_VWRC30143.405 spec_VWRC30143.405
Produktspezifikation
30143 - SULPHURIC ACID 0,25 MOLL 0,5 N AVS TITR _ BSE_rev.0.pdf
BSE/TSE-Zertifikate
SDS_EN_UK_VWRC30143.pdf sds_VWRC30143
Sicherheitsdatenblatt (SDS)
Frequently Bought Together