
Sodium hydroxide 10 mol/l (10 N) in aqueous solution
VWRS5041.9010
:
- Pack type:Plastic container
- Pk:10 L
- Seq:5214
:
:40 g/mol
:109 °C (1013 hPa)
:≤1.11 g/cm³ (20 °C)
:Ambient
:215-185-5
:1824
:8,II
:GHS05
:1310-73-2
:NaOH
:MFCD00003548