

Perchloric acid 0.6 mol/l (0.6 N) in aqueous solution
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7601-90-3
:
Formula:HClO₄
MDL Number:MFCD00011325
:100.46 g/mol
Storage Temperature:Ambient
EINECS:231-512-4
UN:1802
ADR:8,II
:GHS03
:GHS07
Frequently Bought Together