ContactUsLinkComponent
Hydrochloric acid 0.5 mol/l (0.5 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Corrosive
Hydrochloric acid 0.5 mol/l (0.5 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Hydrochloric acid 0.5 mol/l (0.5 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Titer (20°C; real value 0.2 % accuracy):
    0.499 - 0.501 mol/l
  • NIST traceable:
    Confirmed