
Hydrochloric acid 0.357 mol/l (N/2.8), Titripur® volumetric solution, Supelco®
Artikelnr. 1.13136.9010
Supplier: Merck
CAS Number:
Specifications
- Pack type:PE/alu Titripac®
- Pk:10 L
- Seq:5214
Specifications
About this item
:
ADR:8,
:M-Clarity™ quality level = MQ100
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324