Hydrochloric acid 0,1 mol/l (0,1 N), Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®
: Merck
:7647-01-0
:
:HCl
:MFCD00011324
:1.004 g/cm³ (25 °C)
:Ambient
:8,II
:M-Clarity™ quality level = MQ100
:36.46 g/mol
:231-595-7
:1789
:GHS05
- Form:liquid
- Amount-of-substance concentration:0.0995-0.1005
- Measurement uncertainty:+/- 0.0003
- Traceability:NIST SRM