ContactUsLinkComponent
Sulphuric acid concentrate for dilution to 0.5 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Sulphuric acid concentrate for dilution to 0.5 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
  32053.602
 :  
undefined
Sulphuric acid concentrate for dilution to 0.5 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
  32053.602
 :  
  • Pack type:
    Plastic ampoule
  • Pk:
    170 mL
  • Seq:
    2515
  • Titer (20°C):
    0.4975 - 0.5025 mol/l
 :
 :98.08 g/mol
 :8,II
 :Dose for preparation of 1 litre of solution.
 :231-639-5
 :2796
 :GHS05
 :Ambient
 :7664-93-9
 :H₂SO₄
 :MFCD00064589