Order Entry
Germany
ContactUsLinkComponent
 
Flammable
Irritant
DL-Camphor, tablets, TECHNICAL, refined
Catalog # 22478.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
DL-Camphor, tablets, TECHNICAL, refined
Catalog # 22478.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :152.24 g/mol
Boiling Pt:204 °C (1013 hPa)
Melting Pt:∼180 °C
Density:0.992 g/cm³ (25 °C)
Flash Pt:66 °C
Storage Temperature:Ambient
EINECS:200-945-0
UN:2717
ADR:4.1,III
 :GHS02
 :GHS07
CAS Number:76-22-2
Formula:C₁₀H₁₆O
MDL Number:MFCD00074738