(±)-2-Butanol, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:78-92-2
:
Formula:H₃CCH₂CH(OH)CH₃
MDL Number:MFCD00004569
:74.12 g/mol
Merck Index:13,01540
Boiling Pt:94 °C (1013 hPa)
Melting Pt:–115 °C
Density:0.8534 g/cm³ (20 °C)
EINECS:201-158-5
UN:1120
ADR:3,III
:M-Clarity™ quality level = MQ100
Flash Pt:23 °C
:GHS02
:GHS07
Storage Temperature:Ambient