(±)-2-Butanol ≥99%
Catalog # 24126-30KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Stainless steel can
- Pk:30 kg
- Seq:4319
Specifications
Specification Test Results
- Assay (GC):Min. 99 %
- Non-volatile matter:Max. 0.005 %
- Water (Karl Fischer):Max. 0.3 %
Specification Test Results
About this item
:
:74.12 g/mol
Merck Index:13,01540
Boiling Pt:99 °C
Melting Pt:-89 °C
Melting Pt:–89 °C
Density:0.81 g/cm³ (20 °C)
Flash Pt:24 °C
Storage Temperature:Ambient
EINECS:201-158-5
UN:1120
ADR:3,III
:GHS02
:GHS07
CAS Number:78-92-2
Formula:H₃CCH₂CH(OH)CH₃
MDL Number:MFCD00004569