(±)-2-Butanol 99%, extra pure
:78-92-2
:
:H₃CCH₂CH(OH)CH₃
:MFCD00004569
:74.12 g/mol
:13,01540
:99 °C (760 mmHg)
:–115 °C
:0.81 g/cm³
:24 °C
:Ambient
:201-158-5
:1120
:3,III
:GHS07
:GHS02
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3960 to 1.3980 (20°C, 589 nm)
- Water:≤0.5 %
- GC:≥98.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %