0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Osmium tetraoxide
Osmium tetraoxide 4% in aqueous solution
Osmium tetraoxide 4% in aqueous solution
POLS0972A-20
:
POLYSCIENCES
:
Osmium tetraoxide 4% in aqueous solution
POLS0972A-20
:
POLYSCIENCES
:
Pack type:
Glass bottle
Pk:
40 mL
Seq:
2360
:
:
254.23 g/mol
:
Refrigerator
:
244-058-7
:
3287
:
6.1,II
:
GHS06
:
20816-12-0
:
O₄Os
:
MFCD00011150