Sulphuric acid ≥85%
Catalog # OVERSCHWEFEL85
Supplier: OVERLACK CHEMISCHE PRODUKTE
CAS Number:
Specifications
- Pk:100 kg
- Seq:4553
Specifications
About this item
:
:98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589