Order Entry
Germany
ContactUsLinkComponent
Sulphuric acid 50%, TECHNICAL
Corrosive
Sulphuric acid 50%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Sulphuric acid 50%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 50 %

Specification Test Results

Frequently Bought Together