Order Entry
Germany
ContactUsLinkComponent
Orthophosphoric acid ≥85% ACS
Orthophosphoric acid ≥85% ACS
Supplier:  VWR Chemicals
Certificates
undefined
Orthophosphoric acid ≥85% ACS
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Antimony:
    <0.002%
  • APHA:
    ≤10
  • Arsenic:
    ≤0.0001%
  • Assay:
    ≤85%
  • Calcium:
    ≤0.002%
  • Chloride:
    ≤0.0003%
  • Heavy Metals:
    ≤0.001%
  • Insoluble Matter:
    ≤0.001%
  • Iron:
    ≤0.003%
  • Magnesium:
    ≤0.002%
  • Manganese:
    ≤0.5 ppm
  • Nitrate:
    ≤0.0005%
  • Potassium:
    ≤0.005%
  • Reducing Substances:
    Pass
  • Sodium:
    ≤0.025%
  • Sulphate:
    ≤0.003%
  • Volatile Acids:
    ≤0.001%

Specification Test Results

Related Information