Order Entry
Germany
ContactUsLinkComponent
 
 
Orthophosphoric acid ≥75%
  VWRS95021.8000
 :  
 
Orthophosphoric acid ≥75%
  VWRS95021.8000
 :  

 

  • Pack type:
    Plastic container
  • Pk:
    8 L
  • Seq:
    5210

 

 

 :
 :98 g/mol
 :1.580 g/cm³ (20 °C)
 :Ambient
 :231-633-2
 :1805
 :8,III
 :GHS05
 :7664-38-2
 :H₃PO₄
 :MFCD00011340