0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Orthophosphoric acid
Orthophosphoric acid 100 ml/l analytical reagent, for TOC
Orthophosphoric acid 100 ml/l analytical reagent, for TOC
:
Bernd Kraft
Orthophosphoric acid 100 ml/l analytical reagent, for TOC
:
Bernd Kraft
:
7664-38-2
:
:
H₃PO₄
:
MFCD00011340
:
98 g/mol
:
∼1.1 g/cm³ (20 °C)
:
Ambient
:
231-633-2
:
1805
:
8,III
:
GHS07