Order Entry
Germany
ContactUsLinkComponent
Hydrogen peroxide 6% (w/v) (20 volumes), GPR RECTAPUR®
Hydrogen peroxide 6% (w/v) (20 volumes), GPR RECTAPUR®
Catalog # 285175C
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hydrogen peroxide 6% (w/v) (20 volumes), GPR RECTAPUR®
Catalog # 285175C
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2 L
  • Seq:
    5086

Specifications

Specification Test Results

  • Assay (W/V):
    Min. 6 %
  • Density (20/4):
    About 1.020
  • Cl (Chloride):
    Max. 20 ppm
  • Fe (Iron):
    Max. 1 ppm

Specification Test Results

About this item

 :
Density:1.022 g/cm³ (20 °C)
 :34.01 g/mol
EINECS:231-765-0
 :GHS07
Storage Temperature:Ambient
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333