Hydrogen peroxide 50% (w/w) in water stabilised, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt:>= 100 °C Dec. (on heating)
Melting Pt:<0 °C
Density:1.1-1.3 g/cm³ (20 °C)
Flash Pt: Does not flash
Storage Temperature:Ambient
EINECS:231-765-0
UN:2014
ADR:5.1,II
:Stabilised with Stabilised
:34.01 g/mol
:GHS07
:GHS05
:GHS03
Specification Test Results
- Assay:48.0 - 52.0 %
- Vapor Pressure:23.3 mmHg ( 30 °C)