Order Entry
Germany
ContactUsLinkComponent
Hydrogen peroxide 35% stabilised, SAFC®
Hydrogen peroxide 35% stabilised, SAFC®
Supplier:  Merck
Certificates
Hydrogen peroxide 35% stabilised, SAFC®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (manganometric):
    34.5 - 36.0 %
  • Identity:
    passes test
  • Appearance of the substance:
    passes test
  • Free acid (as sulfuric acid):
    ≤ 0.025 %
  • Other heavy metals (as Pb):
    ≤ 0.0002 %
  • Chloride (Cl):
    ≤ 0.005 %
  • Residual solvents (ICH Q3C):
    ...excluded by production process
  • Non volatile matter:
    ≤ 0.10 %
  • Residue on ignition:
    ≤ 0.05 %

Specification Test Results

Related Information